Skip to Content
Merck
All Photos(1)

Documents

AX1440

Supelco

Isoamyl Alcohol

Meets ACS Specifications GR ACS

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-633-5

vapor density

3 (vs air)

Quality Level

vapor pressure

2 mmHg ( 20 °C)

Assay

≥98.5% (corrected for water content, GC)

form

liquid

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

technique(s)

mass spectrometry (MS): suitable

impurities

≤0.002 meq/g Titrable acid
≤0.1% Carbonyl compounds (as HCHO)
≤0.2% Acids and esters (as amyl acetate)
≤0.5% Water (H2O)

evapn. residue

≤0.003%

color

APHA: ≤10

refractive index

n20/D 1.406 (lit.)

pH

7 (20 °C, 25 g/L in H2O)

bp

130 °C (lit.)

mp

−117 °C (lit.)

transition temp

flash point 43.5 °C

solubility

25 g/L

density

0.81 g/cm3 at 20 °C
0.809 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS

Analysis Note

Acids and esters (as amyl acetate)0.2% maxAssay (GC, corrected for water content)98.5% minCarbonyl compounds (as HCHO)0.1% maxColor (APHA)10 maxEvaporation residue0.003% maxFormClear liquidTitrable acid0.002meq/g maxWater (H2O)0.5% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service