Skip to Content
Merck
All Photos(1)

Key Documents

P2069

Sigma-Aldrich

Phenol:Chloroform:Isoamyl Alcohol 25:24:1 Saturated with 10 mM Tris, pH 8.0, 1 mM EDTA

Supplied with Equilibration buffer, for molecular biology

Synonym(s):

DNA extraction

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$145.00
400 ML
$442.00

$145.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details



Select a Size

Change View
100 ML
$145.00
400 ML
$442.00

About This Item

UNSPSC Code:
12190000
NACRES:
NA.52
grade:
Molecular Biology
for molecular biology
application(s):
agriculture

$145.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


grade

Molecular Biology
for molecular biology

Assay

≥99.0%

form

liquid

pH

6.5-6.9(pH of phenolic phase)
7.8-8.2(pH of phenolic phase after the addition of Equilibration Buffer)

application(s)

agriculture

storage temp.

2-8°C

Application

For applications requiring a higher pH, such as the isolation of large intact genomic DNA, addition of the Equilibration Buffer is recommended.
Phenol:Chloroform:Isoamyl Alcohol has been used in the preparation of RNA for the study of total RNA isolation from myocardial biopsies.[1] It has been also used to extract DNA during Human Androgen Receptor (HUMARA) Clonality Assay.[2]

Packaging

Supplied with P3803 and a separate bottle of equilibration buffer to adjust the phenol phase to pH 8.0 ± 0.2.

Suitability

This lot of phenol:chloroform:isoamyl alcohol 25:24:1 was use tested by extracting a crude preparation of pBR322 DNA preparation from E. coli. Based on evaluation by agarose gel electrophoresis the extracted DNA was not degraded or reduced in concentration. The same procedure was applied to purified pBR322 DNA and lambda Hind III digest DNA. Based on electrophoretic evaluation the purified preparations were not detectably degraded or reduced in concentration.

Preparation Note

Add the entire contents of the small bottle of Equilibration Buffer to the large bottle of Phenol:Chloroform:Isoamyl Alcohol. Mix gently and allow the phases to separate before use, approximately 2-4 hours. This will adjust the pH of the Phenol solution from pH 6.7 ± 0.2 to 8.0 ± 0.2.

Other Notes

After the addition of Equilibration Buffer, the product is stable for 6 months vs. 2 years without added buffer. Storage at -20 °C protected from light extends the shelf life to greater than 1 year after the addition of Equilibration Buffer.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongzhong Wang et al.
Microcirculation (New York, N.Y. : 1994), 13(3), 187-197 (2006-04-22)
Angiogenic therapies in animals have demonstrated the development of new blood vessels within ischemic myocardium. However, results from clinical protein and gene angiogenic trials have been less impressive. The present study aimed to investigate the expression of angiogenic genes in
Ellen Marie Straarup et al.
Nucleic acids research, 38(20), 7100-7111 (2010-07-10)
The potency and specificity of locked nucleic acid (LNA) antisense oligonucleotides was investigated as a function of length and affinity. The oligonucleotides were designed to target apolipoprotein B (apoB) and were investigated both in vitro and in vivo. The high
Ole William Petersen et al.
The American journal of pathology, 162(2), 391-402 (2003-01-28)
A breast carcinoma biopsy showed cytochemical evidence of epithelial mesenchymal transition and an alpha-smooth muscle actin-positive stromal reaction. To study the lineage, and the nature of the cells in the stromal reaction, we derived a novel cell line, HBFL-1, from
Amber L Scott et al.
Molecular biology and evolution, 34(10), 2690-2703 (2017-09-29)
Polyploidization events have occurred during the evolution of many fungi, plant, and animal species and are thought to contribute to speciation and tumorigenesis, however little is known about how ploidy level contributes to adaptation at the molecular level. Here we
Martin Häring et al.
Frontiers in behavioral neuroscience, 9, 235-235 (2015-09-22)
The endocannabinoid (eCB) system possesses neuromodulatory functions by influencing the release of various neurotransmitters, including γ-aminobutyric acid (GABA) and glutamate. A functional interaction between eCBs and the serotonergic system has already been suggested. Previously, we showed that cannabinoid type-1 (CB1)

Protocols

Method for reverse transcription of RNA into DNA. Uses a premixed reagent that contains reverse transcriptase, dNTPs, primers, RNase inhibitor and buffer. Fast generation of cDNA.

Questions

1–8 of 8 Questions  
  1. Hello, I used P2069 for DNA extraction from mouse tails. After adding the equilibration buffer, I wait for 2~4hrs until phase seperation. At this point, I have a question. I heard that lower phase is the phase I utilize, so Can I aliquot the lower phase?

    1 answer
    1. The contents of the Tris base are added to the phenol:chloroform:isoamyl alcohol mixture, and the contents are well mixed. The amount added is 6.5 mL equilibration buffer per 100 mL phenol, designed so that only one treatment is necessary to raise the pH. Allow the mixture to separate, which may take 2-4 hours. The upper aqueous phase is removed and discarded. The remaining lower organic phase is divided into aliquots and stored at -20 °C. The solution should be stable for up to 12 months. The pH of the phenol should be raised from 6.7 to about 7.9-8. Only one treatment is needed.

      Helpful?

  2. Hello, I used this material for DNA extraction from mouse tail. After mixing the equilibration buffer, there was two phase. Which phase should I use? Also, Can I aliquot the liquid?

    1 answer
    1. The lower phase should be used for DNA extraction. It is possible to aliquot the liquid immediately after mixing, before the phases separate.

      Helpful?

  3. After equilibrating with the supplied Tris buffer, can I aliquot this for use, if so, should I just top each aliquot with additional Tris pH 8?

    1 answer
    1. Please see the link below to review the product datasheet. Note that the prepration instructions require the addition of the entire vial (6.5 mL) of equilabration buffer to the contents of the product. Aliquot immediately after the gentle mixing step. During the 2 - 4 hour incubation period the mixure will phase separate. At this point the aliquots may be stored for long term use according to the instructions.
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/144/892/p2069dat.pdf

      Helpful?

  4. There is an Equilibration Buffer (Product B5658) that is included with Product P2069, Phenol:Chloroform:Isoamyl Alcohol 25:24:1.  Is B5658 also available separately?

    1 answer
    1. No, it is only available as part of P2069.

      Helpful?

  5. I see that you list two equilibrated Phenol:Chloroform:Isoamyl alcohol products. When should I use Product P3803, and when should I use Product P2069?

    1 answer
    1. In short, P3803 is for RNA work, and P2069 is for DNA work. Product P2069 consists of one bottle of Product P3803 and one bottle of Product B5658. The B5658 is added to the P3803 to raise the pH of the organic phase from 6.7 to 8.0; pH 8.0 is not suitable for RNA work, because RNA can degrade at the higher pH.

      Helpful?

  6. I ordered Product P2069, Phenol:Chloroform:Isoamyl Alcohol 25:24:1; I got one bottle of the Phenol:Chloroform:Isoamyl Alcohol and one bottle of B5658, Equilibration buffer. When and how am I supposed to use the B5658?

    1 answer
    1. B5658 is a Tris buffer that is used to raise the pH of the phenol:chloroform:isoamyl alcohol mixture, to make it suitable for isolation of genomic DNA. The contents of the two bottles need to be well mixed. Then allow the mixture to separate; this may take 2-4 hours. This procedure should raise the pH of the phenol phase from 6.7 to about 8.0.  Only one "treatment" is needed. This solution should be stable for up to 12 months.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. There are two layers in the bottle of Product P2069, Phenol:Chloroform:Isoamyl Alcohol 25:24:1; which layer should I use?

    1 answer
    1. The lower phase is the phase that is to be used.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service