Skip to Content
Merck
All Photos(1)

Documents

AX1308

Supelco

Ammonium Hydroxide

OmniTrace® Ultra

Synonym(s):

Ammonium hydroxide solution, aa, Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352305

grade

for inorganic trace analysis

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 200.7
suitable for EPA 200.8
suitable for EPA 533
suitable for EPA OTM-45
suitable for ISO 21675 2019
suitable for ISO 25101

reg. compliance

suitable for FDA C-010.02

form

liquid

expl. lim.

27 %

manufacturer/tradename

OmniTrace® Ultra

pH

(alkaline)

solubility

soluble (Water: Soluble)

density

0.9 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

OmniTrace Ultra

Application

OmniTrace Ultra
Product is double-distilled and packed in class 100 clean room conditions into preleached HDPE bottles. Each HDPE bottle is packed in an EPS innerpack and then packed into a 1x regulated carton with a Certificate of Analysis.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the separation of several charge variants of two monoclonal antibodies, NISTmAb and Infliximab, using a polymeric cation exchange column and MS-compatible mobile phases.

Glyphosate analysis: LC-MS/MS method with Supel™ Carbon LC U/HPLC column for stability and retention.

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service