1.05428
Ammonia solution
25%, Suprapur®
Synonym(s):
Ammonium hydroxide solution, Ammonia aqueous, Ammonia water
About This Item
Recommended Products
product name
Ammonia solution 25 %, Suprapur®
grade
for inorganic trace analysis
Quality Level
vapor pressure
483 hPa ( 20 °C)
form
liquid
expl. lim.
27 %
concentration
25%
bp
37.7 °C/1013 hPa
mp
-57.5 °C
density
0.9 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
[NH4+].[OH-]
InChI
1S/H3N.H2O/h1H3;1H2
InChI key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc.: This study focuses on the application of 25% ammonia solution in synthesizing zinc-ammonium phosphate from galvanic waste, contributing to waste valorization and chemical recycling efforts (Marian et al., 2024).
Development of a Simultaneous Liquid Chromatography-Tandem Mass Spectrometry Analytical Method for Urinary Endogenous Substrates and Metabolites for Predicting Cytochrome P450 3A4 Activity.: Describes the use of 25% ammonia solution in the preparation phase of samples for advanced chromatographic analyses, crucial for metabolic studies in pharmaceutical research (Mano et al., 2023).
[The study of fenspiride: goals and objectives for chemical and toxicological studies].: Discusses methodologies incorporating 25% ammonia solution in toxicological evaluations, highlighting its role in enhancing detection sensitivities in forensic and clinical toxicology (Barsegyan et al., 2019).
The use of a new, modified Dittmer-Lester spray reagent for phospholipid determination by the TLC image analysis technique.: Investigates the efficacy of 25% ammonia solution as a component of a novel spray reagent for thin-layer chromatography, enhancing lipid analysis capabilities in biochemical studies (Zakrzewski et al., 2013).
Ultrasonic synthesis of stable oil filled microcapsules: This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like ᴅʟ-N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (Source).
Linkage
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
TLC plates, with silica gel, gypsum as binder, International Pharmacopoeia compliant, for identification of Atenolol and Chlorthalidone in tablets.
Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service