Skip to Content
Merck
All Photos(1)

Key Documents

AX0152

Supelco

Acetonitrile

≥99.9% (GC), anhydrous, biotech. grade

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
grade:
anhydrous
biotech. grade
technique(s):
GC/GC: suitable
application(s):
pharmaceutical
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Acetonitrile, anhydrous, Biotechnology, ≥99.9% (GC)

grade

anhydrous
biotech. grade

Quality Level

200
300

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

GC/GC: suitable

impurities

≤0.6 μeq/g Titrable base
≤1 ppm Free amine (as dimethylamine)
≤10 ppm Water (H2O)
≤2.0 μeq/g Titrable acid

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

UV cutoff

≤200 nm

UV absorption

λ: 200 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.60
λ: 220 nm Amax: ≤0.30
λ: 230 nm Amax: ≤0.08
λ: 240 nm Amax: ≤0.03
λ: 250 nm Amax: ≤0.03
λ: 270 nm Amax: ≤0.02
λ: 300 nm Amax: ≤0.01
λ: 350 nm Amax: ≤0.01

application(s)

pharmaceutical

format

neat

shipped in

ambient

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

anhydrous, Biotechnology

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.9% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFree amine (as dimethylamine)1ppm maxRefractive index (n 20/D)1.3434-1.3446Titrable acid2.0µeq/g maxTitrable base0.6µeq/g maxUV Abs. at 200 nm1.00AU maxUV Abs. at 210 nm0.60AU maxUV Abs. at 220 nm0.30AU maxUV Abs. at 230 nm0.08AU maxUV Abs. at 240 nm0.03AU maxUV Abs. at 250 nm0.03AU maxUV Abs. at 270 nm0.02AU maxUV Abs. at 300 nm0.01AU maxUV Abs. at 350 nm0.01AU maxUV Cut-off200nm maxWater (H2O)10ppm max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

The 4L M-Bottle provides safe handling of solvents due to an innovative solvent bottle design with sealing technology to eliminate leaks and reduce risks in solvent dispensing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service