Acetonitrile
Synonym(s):
ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide, Acetonitrile
Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-20 of 41
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3