Skip to Content
Merck
All Photos(2)

Key Documents

ABT66

Sigma-Aldrich

Anti-CADM1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cell adhesion molecule 1, Immunoglobulin superfamily member 4, IgSF4, Nectin-like protein 2, NECL-2, Spermatogenic immunoglobulin superfamily, SgIgSF, Synaptic cell adhesion molecule, SynCAM, Tumor suppressor in lung cancer 1, TSLC-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CADM1(23705)

General description

CADM1 (also known as cell adhesion molecule 1, Nectin-like protein 2, synaptic cell adhesion molecule. IgSF4 and TSLC-1) is a tumor suppressor part of the immunoglobulin superfamily and has been found to show reduced expression in many different solid tumors. CADM1 has also been found to play a role in the formation of epithelia-like cell structure. Recent research has also linked CADM1 to mediating emotional and social behaviors and motor skills. CADM1 may also be important in psychiatric and behavioral disorders such as autism.

Immunogen

Linear peptide corresponding to human CADM1.

Application

Detect CADM1 using this Anti-CADM1 Antibody validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected CADM1 in NIH/3T3, A431, and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in PC3 cell lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected CADM1 in 10 µg of PC3 cell lysate.

Target description

~48 kDa & 37 kDa observed.
Isoform 1 (48 kDa), Isoform 2 (37 kDa).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunju Lee et al.
Cancer, 119(9), 1752-1760 (2013-01-12)
The objective of this study was to discover molecular biomarkers associated with the recurrence of esophageal squamous cell carcinoma (ESCC). The authors retrospectively analyzed the hypermethylation status of 11 genes using methylation-specific polymerase chain reaction (PCR) and the expression of
Fanqiang Kong et al.
Oncology letters, 21(2), 103-103 (2020-12-31)
Glioblastoma (GBM) is a type of malignant tumor occurring in the brain that severely influences the life of affected individuals. GBM cells are highly infiltrative, which is one of the main obstacles in the treatment of the disease. Numerous microRNAs
Peter J Chockley et al.
The Journal of clinical investigation, 128(4), 1384-1396 (2018-01-13)
During epithelial-mesenchymal transition (EMT) epithelial cancer cells transdifferentiate into highly motile, invasive, mesenchymal-like cells, giving rise to disseminating tumor cells. Few of these disseminated cells successfully metastasize. Immune cells and inflammation in the tumor microenvironment were shown to drive EMT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service