Skip to Content
Merck
All Photos(2)

Documents

ABT174

Sigma-Aldrich

Anti-LGN Antibody

serum, from rabbit

Synonym(s):

G-protein-signaling modulator 2, Mosaic protein LGN, LGN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Gpsm2(76123)

General description

LGN is also known as G-protein-signaling modulator 2 (GPSM2). LGN has an important role in cellular structure and spindle pole orientation. LGN also stabilizes the apical complex during neuroblast divisions. LGN defects lead to Chudley-McCullough syndrome as well as a multiple micronuclei phenotype due to chromosomal mis-segregation and cell division errors.

Specificity

Other Homologies: Human (85% sequence homology), Chicken (75% sequence homology).

Immunogen

GST-tagged recombinant protein corresponding to mouse LGN.

Application

Anti-LGN Antibody is a highly specific rabbit polyclonal antibody, that targets LGN & has been tested in western blotting.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LGN in mouse E16 brain tissue lysate.

Quality

Evaluated by Western Blotting in rat E16 heart tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LGN in rat E16 heart tissue lysate.

Target description

~77 kDa observed. Uncharacterized bands may be observed at ~30 kDa, ~60 kDa, and ~70 kDa in some cell lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Heart tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos P Descovich et al.
eLife, 12 (2023-04-06)
Oriented cell divisions balance self-renewal and differentiation in stratified epithelia such as the skin epidermis. During peak epidermal stratification, the distribution of division angles among basal keratinocyte progenitors is bimodal, with planar and perpendicular divisions driving symmetric and asymmetric daughter
Kanako Chishiki et al.
Genes to cells : devoted to molecular & cellular mechanisms, 22(3), 293-309 (2017-02-12)
Correct cyst morphogenesis of epithelial cells requires apical-basal polarization, which is partly regulated by mitotic spindle orientation, a process dependent on the heterotrimeric G protein subunit Gαi and its binding protein LGN. Here, we show that in three-dimensional culture of
Ouranio Anastasiou et al.
Science signaling, 13(620) (2020-02-27)
Correct selection of the cell division axis is important for cell differentiation, tissue and organ morphogenesis, and homeostasis. Both integrins, which mediate interactions with extracellular matrix (ECM) components such as fibronectin, and cadherins, which mediate interactions between cells, are implicated
Par3-mInsc and G?i3 cooperate to promote oriented epidermal cell divisions through LGN.
Williams, SE; Ratliff, LA; Postiglione, MP; Knoblich, JA; Fuchs, E
Nature Cell Biology null
Kendall J Lough et al.
eLife, 8 (2019-12-14)
During organogenesis, precise control of spindle orientation balances proliferation and differentiation. In the developing murine epidermis, planar and perpendicular divisions yield symmetric and asymmetric fate outcomes, respectively. Classically, division axis specification involves centrosome migration and spindle rotation, events occurring early

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service