Skip to Content
Merck
All Photos(1)

Key Documents

C2855

Sigma-Aldrich

Coelenterazine cp

solid

Synonym(s):

CLZN-cp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

form

solid

Quality Level

storage temp.

−20°C

Biochem/physiol Actions

15 times higher luminescence and faster response time than native coelenterazine (Prod. No. C2230).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Inouye et al.
Biochemical and biophysical research communications, 233(2), 349-353 (1997-04-17)
To investigate the use of various luciferases as reporter protein, the substrate specificity of recombinant Renilla luciferase, Oplophorus luciferase and recombinant apoaequorin was examined using 23 kinds of coelenterazine analogues as the substrate. The intensity of luminescence was generally highest
K Teranishi et al.
Analytical biochemistry, 249(1), 37-43 (1997-06-15)
Eleven new coelenterazine analogs containing the 3,7-dihydroimidazo[1,2-alpha]pyrazin-3-one structure were synthesized. The superoxide-triggered chemiluminescence of these compounds was investigated using the hypoxanthine-xanthine oxidase system in comparison with four known compounds. The results showed that an alkyl substitution at the position 5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service