Skip to Content
Merck
All Photos(3)

Documents

ABN1712

Sigma-Aldrich

Anti-NRL (Neural Retina Leucine Zipper)

from rabbit

Synonym(s):

NRL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Nrl(18185)

General description

Neural retina-specific leucine zipper protein (UniProt: P54846; also known as NRL) is encoded by the Nrl gene (Gene ID: 18185) in murine species. NRL is a bZIP transcription factor of the MAF family that regulates the expression of several rod-specific genes, including RHO and phosphodiesterase 6B. NRL is reported to be present in the nuclei of developing and mature rods but not cones. It interacts with the homeodomain protein CRX and synergistically regulates rhodopsin expression. It is also reported to interact with FIZ1 and this interaction can repress transactivation. It consists of a minimal transactivation domain (aa 30-93), a basic motif (aa 159-185), and a leucine-zipper (aa 187-208). The minimal transactivation domain (MTD) is conserved across the MAF family and it may activate transcription by recruiting TBP and associated factors at the promoters of target genes. In developing mouse retina, Nrl expression is reported to precede the appearance of known markers for differentiated rods. The transcriptional activity of NRL is modulated by sumoylation on RHO and NR2E3 promoters. (Ref.: Prabodha, K et al. (2001). J. Biol. Chem 276 (39): 36824-30).

Specificity

This rabbit polyclonal antibody recognizes murine neural retina-specific leucine zipper protein. It iargets an epitope within 108 amino acids from the N-terminal half.

Immunogen

Epitope: unknown
His-tagged recombinant protein fragment corresponding to 108 amino acids from the N-terminal half of murine neural retina-specific leucine zipper protein.

Application

Anti-NRL (Neural Retina Leucine Zipper), Cat. No. ABN1712, is a highly specific rabbit polyclonal antibody that targets Neural retina-specific leucine zipper protein and has been tested for use in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A 1:100-1:1,000 dilution from a representative lot detected NRL (Neural Retina Leucine Zipper) in frozen mouse retina tissue (Courtesy of Dr. Tiansen Li at NIH).

Western Blotting Analysis: A 1:1,000-1:2,000 dilution from a representative lot detected NRL (Neural Retina Leucine Zipper) in mouse retinal tissue lysate (Courtesy of Dr. Tiansen Li at NIH).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse retinal tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NRL (Neural Retina Leucine Zipper) in mouse retinal tissue lysate.

Target description

~31 kDa observed; 26.08 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol.
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service