Skip to Content
Merck
All Photos(1)

Key Documents

AP124A

Sigma-Aldrich

Goat Anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate

Chemicon®, from goat

Synonym(s):

Anti-Mouse IgG Alkaline Phosphatase, Goat Anti-Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Alkaline Phosphatase-conjugated Affinity Purified Goat anti-Mouse IgG (H+L)
Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Specific for mouse IgG, heavy and light chain.

Application

EIA and Western blots: 1:5,000-1:50,000.

Immunohistochemistry: 1:1,000-1:2,000.

Optimal working dilutions must be determined by end user.
Goat anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Mouse IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Linkage

Replaces: AP126A

Physical form

ImmunoAffinity Purified
Lyophilized. Buffer = 0.01 M Tirs HCl, 0.25 M NaCl, pH 8.0 with

15 mg/mL BSA and 0.05% sodium azide.

RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature.

Storage and Stability

this product is stable for several weeks at 2–8°C as an undiluted liquid. DO NOT FREEZE. For

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nigel Aminake Makoah et al.
BMC biotechnology, 24(1), 59-59 (2024-08-28)
Crimean-Congo hemorrhagic fever (CCHF) is a tick-borne zoonotic disease that presents with severe hemorrhagic manifestations and is associated with significant fatality rates. The causative agent, Crimean-Congo Hemorrhagic Fever Virus (CCHFV), is a high-priority pathogen identified by the World Health Organization
Inactivation of Chikungunya virus by 1,5 iodonapthyl azide.
Sharma, A; Gupta, P; Maheshwari, RK
Virology Journal null
Uman-type neurofilament light antibodies are effective reagents for the imaging of neurodegeneration.
Shaw, et al.
Brain communications, 5, fcad067-fcad067 (2023)
Toshihiko Tsukamoto et al.
Cancer, 106(7), 1489-1497 (2006-03-07)
Lysosomal proteases are implicated in cancer progression and metastasis. In the current study, using subtraction cloning for genes that are differentially expressed in metastasis, the authors isolated a clone encoding ceroid lipofuscinosis, neuronal 2 (CLN2), which is a lysosomal serine
Szabolcs Takáts et al.
Autophagy, 11(8), 1209-1215 (2015-06-11)
The HOPS tethering complex facilitates autophagosome-lysosome fusion by binding to Syx17 (Syntaxin 17), the autophagosomal SNARE. Here we show that loss of the core HOPS complex subunit Vps16A enhances autophagosome formation and slows down Drosophila development. Mechanistically, Tor kinase is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service