Skip to Content
Merck
All Photos(1)

Key Documents

AB5612

Sigma-Aldrich

Anti-Citrulline Antibody

serum, Chemicon®

Synonym(s):

CCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

suitability

not suitable for immunohistochemistry (Paraffin)

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Citrulline in glutaraldehyde fixed tissues or cells.



The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:



Compound Cross-reactivity ratio

Citrulline-G-BSA 1

Arginine-G-BSA 1/>100,000

Glutamine-G-BSA 1/>100,000

Ornithine-G-BSA 1/>100,000



Abbreviations:



(G) Glutaraldehyde

(BSA) Bovine Serum Albumin

Immunogen

Citrulline-Glutaraldehyde-BSA

Application

Anti-Citrulline Antibody is a Rabbit Polyclonal Antibody for the detection of Citrulline & has been validated in ELISA & IHC.
Immunohistochemistry: 1:1,000-1:2,500 glutaraldehyde required for reactivity. (0.5%-2.5% is typical; fluorescence detection is not recommended. Blocking is with 3% normal serum with 0.1-0.2% triton X-100 usually. Primary antibody is diluted only in TBS or PBS with out blocking or triton X-100. Incubations are usually 3 hours 37C or 4C overnight. Detection is by ABC or PAP for best results. Not recommended for paraffin embedded tissues.

ELISA: 1:2500-1:5000

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Physical form

Rabbit antiserum. Liquid with 0.05% sodium azide

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte Thålin et al.
Journal of thrombosis and haemostasis : JTH, 18(10), 2732-2743 (2020-07-13)
Recent data propose a diagnostic and prognostic capacity for citrullinated histone H3 (H3Cit), a marker of neutrophil extracellular traps (NETs), in pathologic conditions such as cancer and thrombosis. However, current research is hampered by lack of standardized assays. We aimed
Priyanka Sharma et al.
Molecular cell, 73(1), 84-96 (2018-11-26)
The post-translational modification of key residues at the C-terminal domain of RNA polymerase II (RNAP2-CTD) coordinates transcription, splicing, and RNA processing by modulating its capacity to act as a landing platform for a variety of protein complexes. Here, we identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service