Skip to Content
Merck
All Photos(1)

Key Documents

8.55070

Sigma-Aldrich

HMBA-PEGA resin

Novabiochem®

Synonym(s):

4-Hydroxymethylbenzoic acid PEGA resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641
NACRES:
NA.22

Quality Level

product line

Novabiochem®
PEGA

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-8°C

General description

This support consists of amino PEGA resin derivatized with Sheppard′s base-labile 4-hydroxymethylbenzoic acid linker [1]. For further information, please refer to the product entry for amino PEGA resin(855015). Note: PEGA resins are supplied swollen in ethanol, with 1g dry resin corresponding to approximately 15 mL of swollen resin. As the beads of PEGA resins become very sticky and easily damaged when shrunk or dried, they are best handled in a swollen state. To use, the appropriate amount of swollen resin should be weighed into a reaction vessel and any residual ethanol removed by copious washings with the appropriate solvent or solvent mixture. After peptide assembly, the resin should be washed with DCM and transferred to the cleavage vessel. Excess DCM can be removed under vacuum, and the shrunk resin treated with the TFA cocktail. The characteristics of the swollen resin, its robustness and high permeability enables its use in both batch and continuous flow synthesis apparatus.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] R. C. Sheppard, et al. (1982) Int. J. Peptide Protein Res., 20, 451.

Linkage

Replaces: 01-64-0113

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.10 - 0.40 mmol/g
Swelling Volume (in DMF): lot specific result
dry resin %: lot specific result
The resin is sold swollen in ethanol.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service