Skip to Content
Merck
All Photos(1)

Documents

8.52323

Sigma-Aldrich

Fmoc-homopropargylglycine

Novabiochem®

Synonym(s):

Fmoc-homopropargylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19NO4
CAS Number:
Molecular Weight:
349.38
UNSPSC Code:
12352209

product line

Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alkyne

storage temp.

15-25°C

General description

Building block for the introduction by Fmoc SPPS of an amino acid bearing an alkyne side chain. Such derivatives are useful tools for the synthesis of cyclic peptides by an alkyne-alkyne Glaser coupling, involving Cu(OAc)2/pyridine and heating to 60 °C using microwaves.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally-Protected Building Blocks for Fmoc SPPS

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service