Skip to Content
Merck
All Photos(1)

Key Documents

714291

Sigma-Aldrich

(S)-5-Azido-2-(Fmoc-amino)pentanoic acid

≥97.0% (HPLC), for peptide synthesis

Synonym(s):

Fmoc-5-azido-L-norvaline, Fmoc-L-δ-azidoornithine, Fmoc-Orn(N2)-OH

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$554.00

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$554.00

About This Item

Empirical Formula (Hill Notation):
C20H20N4O4
CAS Number:
Molecular Weight:
380.40
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

(S)-5-Azido-2-(Fmoc-amino)pentanoic acid, ≥97.0% (HPLC)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]/D 3.5±0.3°, c = 1 in ethyl acetate

composition

carbon, 62.2-63.9%
nitrogen, 14.3-15.1%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reaction type: click chemistry

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CCCN=[N+]=[N-])NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C20H20N4O4/c21-24-22-11-5-10-18(19(25)26)23-20(27)28-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2,(H,23,27)(H,25,26)/t18-/m0/s1

InChI key

TVPIDQLSARDIPX-SFHVURJKSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucio Litti et al.
Nanoscale, 10(3), 1272-1278 (2018-01-03)
Multimodal contrast agents offer new interesting diagnostic possibilities, summing the benefits of multiple imaging techniques. Magnetic resonance and optical imaging are complementary techniques. The first allows total body screening, even though it suffers from low spatial resolution and needs high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service