8.51004
BOP
Benzotriazole-1-yl-oxy-tris-(dimethylamino)-phosphonium hexafluorophosphate Novabiochem®
Synonym(s):
BOP, Benzotriazole-1-yl-oxy-tris-(dimethylamino)-phosphoniumhexafluorophosphate, Castro′s Reagent
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
Assay
≥99.0% (HPLC)
form
powder
reaction suitability
reaction type: Coupling Reactions
manufacturer/tradename
Novabiochem®
mp
138-145 °C
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
F[P-](F)(F)(F)(F)F.[P](=[N+](C)C)(N(C)C)(N(C)C)O[n]1nnc2c1cccc2
InChI
1S/C12H22N6OP.F6P/c1-15(2)20(16(3)4,17(5)6)19-18-12-10-8-7-9-11(12)13-14-18;1-7(2,3,4,5)6/h7-10H,1-6H3;/q+1;-1
InChI key
MGEVGECQZUIPSV-UHFFFAOYSA-N
General description
Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents Literature references
[1] B. Castro, et al. (1975) Tetrahedron Lett., 16, 1219.
[2] B. Castro, et al. (1976) Synthesis, 751.
[3] B. Castro, et al. (1977) Synthesis, 413.
[4] B. Castro, et al. (1977) J. Chem. Res. (S), 182.
[5] P. Rivaille, et al. (1980) Tetrahedron, 36, 3413.
[6] D. L. Nguyen, et al. (1985) J. Chem. Soc., Perkin Trans. 1, 1025.
[7] D. L. Nguyen, et al. (1987) J. Chem. Soc., Perkin Trans. 1, 1915.
[8] A. Fournier, et al. (1988) Int. J. Peptide Protein Res., 31, 86 and 231.
[9] J.-P. Briand, et al. (1989) Pept. Res., 2, 381.
[10] A. Fournier, et al. (1989) Int. J. Peptide Protein Res., 33, 133.
[11] D. L. Nguyen, et al. (1989) Biochem. Biophys. Res. Commun., 162, 1425.
[12] W. K. Rule, et al. in ′Peptides 1988, Proc. 20th European Peptide Symposium′, G. Jung & E. Bayer (Eds), Walter de Gruyter, Berlin, 1989, pp. 238.
[13] M. Forest, et al. (1990) Int. J. Peptide Protein Res., 35, 89.
[14] R. Seyer, et al. (1990) Int. J. Peptide Protein Res., 35, 465.
[15] H. Gausepohl, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 2nd International Symposium′, R. Epton (Eds), Intercept UK Ltd., Andover, 1993, pp. 387.
[16] R. P. McGeary (1998) Tetrahedron Lett., 39, 3319.
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (0.6 mmole in 1 ml DMF): clearly soluble
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.1A - Other explosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service