Skip to Content
Merck
All Photos(3)

Key Documents

712191

Sigma-Aldrich

COMU®

greener alternative

≥96.5% (TLC), for peptide synthesis

Synonym(s):

(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholino-carbenium hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$100.00
25 G
$361.00
100 G
$956.00

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$100.00
25 G
$361.00
100 G
$956.00

About This Item

Empirical Formula (Hill Notation):
C12H19F6N4O4P
CAS Number:
Molecular Weight:
428.27
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

COMU®, 97%

Quality Level

Assay

≥96.5% (TLC)
96.5-103.5% (T)
97%

form

crystals

reaction suitability

reaction type: Coupling Reactions

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

peptide synthesis

functional group

amine
ester
ether
nitrile

greener alternative category

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NO\C(N1CCOCC1)=[N+](/C)C)C#N

InChI

1S/C12H19N4O4.F6P/c1-4-19-11(17)10(9-13)14-20-12(15(2)3)16-5-7-18-8-6-16;1-7(2,3,4,5)6/h4-8H2,1-3H3;/q+1;-1/b14-10-;

InChI key

GPDHNZNLPKYHCN-DZOOLQPHSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Safer, non-explosive and more effective replacement for Benzotriazole-Based peptide coupling reagents and thus aligns with "Less Hazardous Chemical Synthesis" and "Inherently Safer Chemistry for Accident Prevention". Click here for more information.

Application

COMU – Safer and More Efficient Peptide Coupling Reagent

Advantages
  • Equal or even superior performance to HATU
  • Non-explosive (does not contain benzotriazole moiety)
  • Suitable for solution phase & solid phase peptide synthesis
  • Utmost retention of configuration – low to non-existent racemization
  • High solubility and stability in typical solvents
  • Visual or colorimetric reaction monitoring possible
  • Easy removal of water-soluble by-products

Legal Information

COMU is a registered trademark of Luxembourg Bio Technologies Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service