Skip to Content
Merck
All Photos(1)

Key Documents

8.22133

Sigma-Aldrich

Propionaldehyde

for synthesis

Synonym(s):

Propionaldehyde, Propanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-623-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

343 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

175 °C

potency

1410 mg/kg LD50, oral (Rat)
2460 mg/kg LD50, skin (Rabbit)

expl. lim.

2.3-21 % (v/v)

bp

47-48 °C/1013 hPa

mp

-81 °C

transition temp

flash point -40 °C

density

0.81 g/cm3 at 20 °C

InChI

1S/C3H6O/c1-2-3-4/h3H,2H2,1H3

InChI key

NBBJYMSMWIIQGU-UHFFFAOYSA-N

Application


  • Propanethiol Oxidation Pathway: Propionaldehyde is a critical intermediary in the microbial catabolism of propanethiol, which is essential for environmental bioremediation strategies using Pseudomonas putida. This application in biotechnology suggests promising approaches to industrial waste treatment (Qiao et al., 2024).

  • Chemical Synthesis of mRNA Probes: Propionaldehyde serves as a reagent in the synthesis of modified guanosine phosphate derivatives, which are vital for mRNA labeling in biochemistry and pharmaceutical applications, highlighting its role in advanced molecular biology techniques (Senthilvelan et al., 2023).

  • VOC Treatment in Environmental Engineering: In environmental engineering, propionaldehyde is analyzed as a volatile organic compound (VOC) from service station emissions, underlining its impact on air quality and secondary pollution (Hu et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.797 - 0.802
Water (GC): ≤ 2.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Zamani Ghaleshahi et al.
Food research international (Ottawa, Ont.), 130, 108966-108966 (2020-03-12)
In this work, coated liposome as a novel delivery system assembled from genipin-crosslinked with whey protein isolate (WPI) and sodium alginate (SA) as the second layer on WPI-coated liposomal was developed in order to increase its bioavailability and prolong release

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service