Skip to Content
Merck
All Photos(1)

Documents

8.18872

Sigma-Aldrich

2-Methoxyethylamine

for synthesis

Synonym(s):

2-Methoxyethylamine, 2-Aminoethyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO
CAS Number:
Molecular Weight:
75.11
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-712-1
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

285 °C

potency

1570 mg/kg LD50, oral (Rat)

expl. lim.

3-16.10 % (v/v)

refractive index

n20/589 1.4594

pH

12 (20 °C, 100 g/L in H2O)

bp

95 °C/1013 hPa

mp

-82 °C

transition temp

flash point 12 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.872 - 0.874
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service