Skip to Content
Merck
All Photos(1)

Key Documents

153583

Sigma-Aldrich

1-(Trimethylsilyl)imidazole

96%

Synonym(s):

N-Trimethylsilylimidazole, TMSI, TSIM

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$62.50
25 ML
$166.00
100 ML
$356.00

$62.50


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$62.50
25 ML
$166.00
100 ML
$356.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
Beilstein:
606148
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$62.50


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

synthesis grade

Assay

96%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

93-94 °C/14 mmHg (lit.)

density

0.956 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)n1ccnc1

InChI

1S/C6H12N2Si/c1-9(2,3)8-5-4-7-6-8/h4-6H,1-3H3

InChI key

YKFRUJSEPGHZFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(Trimethylsilyl)imidazole (TMSI) was used for derivatization of carbohydrates into trimethylsilyl ethers[1]. It was also used to synthesize polysubstituted chiral spirotetrahydropyrans[2] and as silylating reagent for the protection of hydroxyl groups in the presence of amine functionalities.[3]
Silylating reagent for the protection of hydroxyl groups in the presence of amine functionalities.[3]

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maiwenn Jacolot et al.
Organic letters, 14(1), 58-61 (2011-12-02)
We report a TMSI-promoted Prins cyclization reaction with ketones as carbonyl partners to prepare polysubstituted chiral spirotetrahydropyrans. In the presence of racemic 2-methylcyclohexanone a dynamic kinetic resolution occurred affording one stereoisomer. The observed enantiospecificity has been rationalized by DFT calculation.
Synthetic Communications, 23, 2191-2191 (1993)
Marijana M Ačanski et al.
Food chemistry, 145, 743-748 (2013-10-17)
Gas chromatography with mass spectrometry was used for carrying out a qualitative analysis of the ethanol soluble flour extract of different types of cereals bread wheat and spelt and pseudocereals (amaranth and buckwheat). TMSI (trimethylsilylimidazole) was used as a reagent
Silylated N,O-ketals from the reaction of ketones with N-trimethylsilylimidazole.
P A Andrews et al.
Journal of chromatography, 419, 271-274 (1987-08-07)
Qin Zhao et al.
Journal of chromatography. A, 1240, 45-51 (2012-04-24)
In the present work, we developed a novel dispersive microextraction technique by combining the advantages of liquid-phase microextraction (LPME) and magnetic solid-phase extraction (MSPE). In this method, trace amount of water directly absorbed on bare Fe₃O₄ to form water-coated Fe₃O₄

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service