8.18713
Methyl nicotinate
for synthesis
Synonym(s):
Methyl nicotinate
About This Item
Recommended Products
vapor pressure
0.4 hPa ( 25 °C)
Quality Level
Assay
≥99.0% (GC)
form
crystals
potency
>2000 mg/kg LD50, oral (Rat)
bp
209 °C/1013 hPa
mp
39 °C
transition temp
flash point 95 °C
bulk density
600 kg/m3
storage temp.
2-30°C
SMILES string
n1cc(ccc1)C(=O)OC
InChI
1S/C7H7NO2/c1-10-7(9)6-3-2-4-8-5-6/h2-5H,1H3
InChI key
YNBADRVTZLEFNH-UHFFFAOYSA-N
Application
- Green synthesis of nicotinamide derivatives: Development of a green synthesis process for nicotinamide derivatives using Novozym® 435 in microreactors demonstrated an innovative application of methyl nicotinate in sustainable chemical production (Sheng et al., 2024).
Analysis Note
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 42 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service