Skip to Content
Merck

Skip To

1.00034

1,1,2,2-Tetrabromoethane

1,1,2,2-Tetrabromoethane. CAS 79-27-6, chemical formula Br2CHCHBr2.

Synonym(s):

1,1,2,2-Tetrabromoethane, TBE, Muthmanns liquid, Acetylene tetrabromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H2Br4
CAS Number:
Molecular Weight:
345.65
UNSPSC Code:
12352101
EC Index Number:
201-191-5
NACRES:
NA.22
MDL number:
Assay:
≥98% (GC)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.03-0.13 hPa ( 20 °C)

Quality Segment

assay

≥98% (GC)

form

liquid

autoignition temp.

335 °C

potency

5250 mg/kg LD50, skin (Rat)

bp

239 °C/1013 hPa (decomposition)

mp

-1 °C

density

2.96 g/cm3 at 25 °C

storage temp.

15-25°C

SMILES string

BrC(Br)C(Br)Br

InChI

1S/C2H2Br4/c3-1(4)2(5)6/h1-2H

InChI key

QXSZNDIIPUOQMB-UHFFFAOYSA-N

Application

  • Tetrabromoethane as ς-Hole Donor toward Bromide Ligands: Halogen Bonding between C2H2Br4 and Bromide Dialkylcyanamide Platinum(II) Complexes: The study investigates XBs with 1,1,2,2-tetrabromoethane as XB donor in the formation of 1:1 complex of C2H2Br4 and bromide dialkylcyanamide Platinum(II) (AM Cheranyova, DM Ivanov, 2021).

Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 2.950 - 2.970
Water (K. F.): ≤ 0.05 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 1,1,2,2-Tetrabromoethane


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
100034100004022536001005

Questions

Reviews

No rating value

Active Filters