Skip to Content
Merck
All Photos(1)

Key Documents

8.18242

Sigma-Aldrich

Oxalic acid dihydrate

for synthesis

Synonym(s):

Oxalic acid dihydrate, Ethanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOH)2 · 2 H2O
CAS Number:
Molecular Weight:
126.07
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-634-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.000312 hPa ( 25 °C)

Quality Level

Assay

≥99% (manganometric)

form

solid

potency

375 mg/kg LD50, oral (Rat)

pH

1.5 (10 g/L in H2O)

bp

149-160 °C/1013 hPa (decomposition)

mp

98-100 °C

transition temp

flash point 157 °C (decomposition)

solubility

>100 g/L

density

1.65 g/cm3 at 20 °C

bulk density

813 kg/m3

storage temp.

2-30°C

SMILES string

[O+H3].[O+H3].[O-]C(=O)C(=O)[O-]

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Related Categories

Application

<ul>
<li><strong>Oxalic Acid Dihydrate in Nanomaterial Synthesis:</strong> During the co-precursor driven solid-state thermal reactions oxalic acid dihydrate is used to produce hematite nanomaterials, providing insights into its role in enhancing reaction kinetics for material science applications (Chakraborty et al., 2023).</li>
<li><strong>Oxalic Acid Dihydrate in Polymer Research:</strong>oxalic acid dihydrate is used in modifying the supramolecular structure of bamboo to facilitate the production of lignin-containing nanocellulose, highlighting its utility in sustainable material processing (Wang et al., 2023).</li>
</ul>

Analysis Note

Assay (manganometric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service