Skip to Content
Merck
All Photos(1)

Documents

1.00492

Supelco

Oxalic acid dihydrate

EMPLURA®

Synonym(s):

Oxalic acid dihydrate, Ethanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOH)2 · 2 H2O
CAS Number:
Molecular Weight:
126.07
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-634-3
NACRES:
NA.21

vapor pressure

0.000312 hPa ( 25 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (manganometric)

form

solid

potency

375 mg/kg LD50, oral (Rat)

ign. residue

≤0.05% (800 °C as sulfate)

pH

1.5 (10 g/L in H2O)

bp

149-160 °C/1013 hPa (decomposition)

mp

98-100 °C

transition temp

flash point 157 °C (decomposition)

density

1.65 g/cm3 at 20 °C

bulk density

813 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.001%
N: ≤0.005%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Related Categories

Application


  • In Situ Investigation of a Self-Accelerated Cocrystal Formation by Grinding Pyrazinamide with Oxalic Acid.: This study investigates the in situ cocrystal formation of pyrazinamide and oxalic acid dihydrate, emphasizing the self-accelerating nature of the process and its implications for pharmaceutical development (Kulla et al., 2016).

  • Deuterium MAS NMR studies of dynamics on multiple timescales: histidine and oxalic acid.: This article examines the dynamics of oxalic acid dihydrate and histidine using deuterium magnetic angle spinning nuclear magnetic resonance, providing insights into molecular motions on various timescales (Chan-Huot et al., 2015).

Analysis Note

Assay (manganometric): ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.02 %
Heavy metals (as Pb): ≤ 0.002 %
Total nitrogen (N): ≤ 0.005 %
Fe (Iron): ≤ 0.001 %
Residue on ignition (800 °C as sulfate): ≤ 0.05 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service