Skip to Content
Merck
All Photos(1)

Key Documents

8.15052

Sigma-Aldrich

Methanol-D3

deuteration degree min. 99.5% for NMR spectroscopy MagniSolv

Synonym(s):

Methanol-D3, Tetradeuteromethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHD3O
CAS Number:
Molecular Weight:
35.06
MDL number:
UNSPSC Code:
12142201
EC Index Number:
217-435-9
Pricing and availability is not currently available.

vapor pressure

129 hPa ( 20 °C)

form

liquid

autoignition temp.

440 °C

potency

5628 mg/kg LD50, oral (Rat)

expl. lim.

6-50 % (v/v)

technique(s)

NMR: suitable

bp

65 °C/1013 hPa

mp

-98 °C

transition temp

flash point 9 °C

density

0.867 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/CH4O/c1-2/h2H,1H3/i1D3

InChI key

OKKJLVBELUTLKV-FIBGUPNXSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.5 %

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service