Skip to Content
Merck
All Photos(2)

Documents

8.14761

Sigma-Aldrich

Tetrakis(triphenylphosphine)-palladium(0)

for synthesis

Synonym(s):

Tetrakis(triphenylphosphine)-palladium(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(P(C6H5)3)4
CAS Number:
Molecular Weight:
1155.56
MDL number:
UNSPSC Code:
12352103
EC Index Number:
238-086-9
NACRES:
NA.22

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

103-107 °C

storage temp.

2-8°C

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

General description

Tetrakis(triphenylphosphine)-palladium(0) can be used to catalyze the formation of carbon-carbon bonds, the reduction of functional groups, the formation of carbon-metal bonds, and the deprotection of allyloxycarbonyl groups.

Application

  • Preparation of tricationic tris(pyridylpalladium(II)) metallacyclophane as an anion receptor.: This research explores the synthesis of a tricationic tris(pyridylpalladium(II)) metallacyclophane, showcasing its potential as an anion receptor. The study emphasizes the versatility of palladium complexes in developing new molecular structures for various applications (Danjo et al., 2022).
  • A Ligand-Enabled Palladium-Catalyzed Highly para-Selective Difluoromethylation of Aromatic Ketones.: This study demonstrates the use of a palladium catalyst for highly selective difluoromethylation of aromatic ketones, highlighting the catalyst′s efficiency in organic synthesis and its potential for developing pharmaceuticals (Tu et al., 2018).
  • Viable pathways for the oxidative addition of iodobenzene to palladium(0)-triphenylphosphine-carbonyl complexes: a theoretical study.: This theoretical investigation explores the oxidative addition mechanisms of iodobenzene to palladium(0)-triphenylphosphine-carbonyl complexes, providing insights into the reactivity and applications of palladium catalysts in organic synthesis (Pálinkás et al., 2017).
  • Morita-Baylis-Hillman Reaction of α,β-Unsaturated Ketones with Allylic Acetates by the Combination of Transition-Metal Catalysis and Organomediation.: The paper discusses a novel Morita-Baylis-Hillman reaction facilitated by palladium(0) catalysts, demonstrating its utility in constructing complex molecular architectures for pharmaceutical synthesis (Li et al., 2016).
  • Synthesis of a sterically crowded atropisomeric 1,8-diacridylnaphthalene for dual-mode enantioselective fluorosensing.: This research highlights the synthesis of a novel atropisomeric compound using palladium(0) catalysts, focusing on its application in enantioselective fluorosensing, crucial for biochemical analyses (Mei et al., 2006).

Analysis Note

Assay (ICP-OES): ≥ 97.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrakis (triphenylphosphine) palladium (0)
Friesen R, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis, 1-7 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service