Skip to Content
Merck
All Photos(1)

Documents

8.06189

Sigma-Aldrich

Malononitrile

for synthesis

Synonym(s):

Malononitrile, Malonodinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2N2
CAS Number:
Molecular Weight:
66.06
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-703-2
NACRES:
NA.22

vapor pressure

1 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

365 °C

potency

350 mg/kg LD50, skin (Rat)

bp

220 °C/1013 hPa

mp

32-34 °C

transition temp

flash point 112 °C

solubility

133 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H2N2/c4-2-1-3-5/h1H2

InChI key

CUONGYYJJVDODC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 30 °C
Melting range (upper value): ≤ 33 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service