Skip to Content
Merck
All Photos(1)

Key Documents

8.01977

Sigma-Aldrich

Butyryl chloride

for synthesis

Synonym(s):

Butyryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)2COCl
CAS Number:
Molecular Weight:
106.55
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-498-5
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

39 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

280 °C

potency

>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.5-7.3 % (v/v)

refractive index

n20/589 1.412

bp

101-105 °C/1013 hPa

mp

-89 °C

transition temp

flash point 18 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CCC

InChI

1S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3

InChI key

DVECBJCOGJRVPX-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.017 - 1.019
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F - DIN 51755 Part 1

Flash Point(C)

18 °C - DIN 51755 Part 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service