Skip to Content
Merck
All Photos(1)

Documents

8.01276

Sigma-Aldrich

Pivaloyl chloride

for synthesis

Synonym(s):

Pivaloyl chloride, 2,2-Dimethylpropionyl chloride, Trimethylacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCl
CAS Number:
Molecular Weight:
120.58
MDL number:
UNSPSC Code:
12352106
EC Index Number:
221-921-6

vapor pressure

39 hPa ( 20 °C)

form

liquid

autoignition temp.

455 °C

potency

1420 mg/kg LD50, oral (Rat)
>2010 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-7.4 % (v/v)

bp

105 °C/1013 hPa

mp

-57 °C

transition temp

flash point 13 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H9ClO/c1-5(2,3)4(6)7/h1-3H3

InChI key

JVSFQJZRHXAUGT-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 0.979 - 0.982
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service