The MVP ICON Temperature Probe is a thermistor housed in a stainless steel probe designed for general purpose temperature measurement of liquids or semi-solids ranging from -55 to 125 °C.
Application
Temperature control can be a critical factor in food safety and temperature must be accurately measured and recorded in many HACCP plans. Temperature is one of the most important factors permitting growth of bacteria and limits can be set for various areas of retail food production.
Features and Benefits
Versatile The MVP ICON Temperature Probe accurately reads temperature across a wide range of applications.
Durable The rugged, detachable temperature probe makes in-plant testing easy. The stainless steel probe features a pierced tip, allowing for easy penetration into food samples.
Components
One MVP ICON Temperature Probe
Compatibility
Any other combination might cause loss of performance or irreversible damage to both probe and instrument.
Suitability
Food Processing, HACCP Monitoring, Sanitation
Testing Method
Temperature
Storage and Stability
Clean probe thoroughly with water and a mild detergent solution. Rinse with deionized water. Store probe in original packaging when not in use.
System Requirements
Windows 7 or Windows 10
Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
At least 16 GB of hard disk space
WXGA video resolution of 1280 x 800 or better
Mouse or other pointing device
One available USB port
Adobe Acrobat Reader or similar PDF Viewer
Printer (optional)
Legal Information
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.