Skip to Content
Merck
All Photos(1)

Key Documents

73006BC

Millipore

GDS Resuspension Plates

BioControl, For use with GDS

Synonym(s):

96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$555.00

$555.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
1 UNIT
$555.00

About This Item

UNSPSC Code:
41104017

$555.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

packaging

box of 120 plates

manufacturer/tradename

BioControl Systems 73006

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

Related Categories

General description

GDS Resuspension Plates are the consumable accessories of the Assurance® GDS assays. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. As part of the sample preparation for Assurance® GDS assays, the Resuspension Plates are used as part of the final step in sample preparation whereby sample magnetic particles are resuspended prior to PCR analysis.

Application

The Assurance® GDS Resuspension Plates are used during sample preparation to hold sample prior to PCR analysis for the detection of food pathogens by Assurance® GDS.

Components

Box of 120 plates

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service