Skip to Content
Merck
All Photos(1)

Key Documents

71091

Sigma-Aldrich

Nova Taq Hot Start DNA Polymerase

Heat-activatable modified form of recombinant Taq DNA polymerase

Synonym(s):

Hot-start Taq DNA polymerase, NovaTaq HS DNA polymerase, hot start PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106314
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bacterial (Thermus aquaticus)

Quality Level

recombinant

expressed in E. coli

form

liquid

usage

sufficient for 1000 reactions
sufficient for 200 reactions

feature

dNTPs included: no
hotstart

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

PCR: suitable

input

purified DNA

suitability

suitable for PCR

shipped in

wet ice

storage temp.

−20°C

General description

NovaTaq Hot Start DNA Polymerase is a chemically modified Taq DNA Polymerase, which is inactive at room temperature. This polymerase is a derived recombinant form of Thermus aquaticus DNA polymerase. The enzyme exhibits a 5′→3′ DNA polymerase activity and lacks 3′→5′ exonuclease activity.[1] The enzyme provides improved specificity when compared to standard Taq polymerase and can eliminate the presence of nonspecific amplification such as primer-dimers and mis-primed products. The enzyme must be activated by heat treatment (7–10 min at 95°C), after which thermal cycling can proceed. NovaTaq Hot Start DNA Polymerase generates PCR products with 3′-dA overhangs, suitable for cloning with Novagen® Perfectly Blunt®, Acceptor, and LIC Vector Kits.[1] NovaTaq Hot Start DNA Polymerase is a component of NovaTaq Hot Start Master Mix Kit.

Source: Recombinant Thermus aquaticus DNA polymerase expressed in E. coli
Concentration: 5.0 U/μl
Endonuclease:None detected
Exonuclease: None detected
Amplification efficiency: Functional PCR
Storage: –20°

Application

NovaTaq Hot Start DNA Polymerase has been used in polymerase chain reaction (PCR) amplification.[2] It is suitable for use in Hot Start routine PCR.

Features and Benefits

  • Higher PCR specificity and yield
  • Improved low-copy target amplification
  • Ambient temperature setup compatible with automation
  • Target amplification of up to 5 kbp
  • Ideal for quantitative and high-throughput PCR applications

Components

•250 U or 5 × 250 UNovaTaq Hot Start DNA Polymerase (5 U/µl)

•1.5 ml or 5 × 1.5 ml10X NovaTaq Hot Start Buffer•1.5 ml or 5 × 1.5 ml25 mM MgCl₂

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the incorporation of 10 nmol of dNTP into acid-insoluble form in 30 min at 72°C, in a reaction containing 25 mM TAPS (tris-[hydroxymethyl]-methyl-amino-propane-sulfonic acid, sodium salt), pH 9.3 at 25°C, 50 mM KCl, 2 mM MgCl₂, 1 mM 2-mercaptoethanol, 0.2 mM dATP, dGTP, and dTTP, 0.1 µM [α-³²P]dCTP, and 12.5 µg activated salmon sperm DNA in a volume of 50 µl.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patents rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California, 94404, USA.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Perfectly Blunt is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Chen et al.
Environmental health perspectives, 118(11), 1597-1602 (2010-06-22)
Evidence of potent rodent carcinogenicity via an unclear mechanism suggests that furan in various foods [leading to an intake of up to 3.5 microg/kg body weight (bw)/day] may present a potential risk to human health. We tested the hypothesis that
Yubao Wang et al.
iScience, 9, 149-160 (2018-11-06)
The role of maternal and embryonic leucine zipper kinase (MELK) in cancer cell proliferation has been contentious, with recent studies arriving at disparate conclusions. We investigated the in vitro dependency of cancer cells on MELK under a range of assay conditions.
Quorum Quenching Effect of Recombinant Paraoxonase-1 Enzyme against Quorum Sensing Genes Produced from Pseudomonas aeruginosa.
Faisal A, et al.
Gene Reports, 101412-101412 (2021)
Factors affecting quantitative PCR assay of Plesiomonas shigelloides.
Gu W and Levin R E
Food Biotechnology, 20, 219-230 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service