Skip to Content
Merck

Skip To

70751

BugBuster® Ni-NTA His•Bind® Purification Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
41106500
Shipped in:
wet ice
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

General description

BugBuster®Ni-NTA HIS-BIND® Purification Kit is used for protein purification. Ni-NTA HIS-BIND® Resin is high-performance Ni2+-charged agarose used for rapid, one-step purification of proteins containing a polyhistidine tag sequence.

Application

BugBuster®Ni-NTA HIS-BIND® Purification Kit has been used for the purification of His tagged proteins such as trehalose-6-phosphate phosphatase (TPP) [1], Ras(WT) proteins[2] and CsgA (major curlin subunit) proteins.[3]

Other Notes

•2 × 100 mlBugBuster Protein Extraction Reagent

•10,000 UBenzonase Nuclease, purity >90%

•10 mlNi-NTA His•Bind Resin

•pkg/4Chromatography Columns
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS-BIND is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
707937089970794
manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

shipped in

-


pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids

wgk

WGK 3

Hazard Classifications

Flam. Liq. 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters