Skip to Content
Merck
All Photos(1)

Key Documents

7052X

Sigma-Aldrich

Papanicolaou OG-6

HARLECO® Cytoplasmic counterstain for Pap staining technique

Synonym(s):

PAP Counterstain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116113

Quality Level

form

liquid

mp

-0.1 °C

transition temp

flash point 15 °C

density

0.79 g/cm3

λmax

493-503 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

Application

HARLECO® Cytoplasmic counterstain for Pap staining technique
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Cytoplasmic counterstain for Pap staining technique.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service