Skip to Content
Merck
All Photos(1)

Key Documents

65044A

Sigma-Aldrich

Hemacolor® Solution I

HARLECO® Fixative for fast staining in hematology and cytology

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
32.04
MDL number:
UNSPSC Code:
41116121
EC Index Number:
200-659-6
application(s):
diagnostic assay manufacturing
hematology
histology
bp:
64.5 °C/1013 hPa
vapor pressure:
128 hPa ( 20 °C)

vapor pressure

128 hPa ( 20 °C)

Quality Level

form

liquid

bp

64.5 °C/1013 hPa

mp

-98 °C

transition temp

flash point 9.7 °C

density

0.792 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

SMILES string

OC

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Application

HARLECO® Fixative for fast staining in hematology and cytology
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
May also be used as a fixative for all Wright and Wright-Giemsa staining protocols.

Components

Replacement for methanol fixative component of the Hemacolor® Stain Set.

Analysis Note

Evaporation Number: 1.9

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. This product was discontinued. The website lists 1.11955 Hemacolor® Rapid staining of blood smear as a similar product. Need a replacement for 65044A. Are these products equivalent?

    1 answer
    1. Yes, product 111955 is equivalent to the discontinued Hemacolor reagent 65044A. However, there are slight differences between the products. In the Harleco products, the two stain solutions are buffered, whereas in product 111955, the two stain solutions do not appear to be buffered. After dipping the slides in the two stain solutions, the slides must then be dipped in a buffer solution. This adds only slightly more time to the procedure, but it does require one additional reagent.

      While 65044A was listed as a 4-liter container, 1119552500 is only a 2.5-liter container.

      Helpful?

  2. is the fixative (catalog# 65044A) solution in Wright stain set 100% methanol?

    1 answer
    1. Yes, this product is essentially 100 % Methanol. There may be trace amounts of water, however, this is not quantitated and does not effect performance.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service