Skip to Content
Merck
All Photos(1)

Key Documents

614849

Sigma-Aldrich

TOPK/PBK Inhibitor, HI-TOPK-032

A cell-permeable, selective, ATP competitive, reversible inhibitor of T-LAK-cell-originated protein kinase (IC₅₀ = 2 µM).

Synonym(s):

TOPK/PBK Inhibitor, HI-TOPK-032

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N5OS
CAS Number:
Molecular Weight:
369.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

rust

solubility

DMSO: 2.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)C(=O)Nc2cc3[n](c4nc5c(nc4c3C#N)cccc5)cc2

InChI

1S/C20H11N5OS/c21-11-13-16-10-12(22-20(26)17-6-3-9-27-17)7-8-25(16)19-18(13)23-14-4-1-2-5-15(14)24-19/h1-10H,(H,22,26)

InChI key

BCSBXWKRZUPFHW-UHFFFAOYSA-N

General description

A cell-permeable, ATP competitive, reversible indolizinoquinoxalin derivative that acts as a specific inhibitor of T-LAK-cell-originated protein kinase (TOPK/PBK)(IC50 = 2 µM). Does not affect the activities of ERK1, JNK1, and p38 MAP kinases. At higher concentrations (~ 5 µM) also inhibits MEK1 activity. Shown to suppress the proliferation of HCT116 colon cancer cell line and block tumor growth in colon cancer xenograft models in a dose-dependent manner. Diminishes TOPK activity only in cells over-expressing TOPK without significantly affecting cells with low levels of expression. Also shown to increase the expression of p53, active caspase-7, cleaved poly-ADP-Ribose polymerase, and increase the phosphorylation of Cdc2.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable, selective, ATP competitive, reversible inhibitor of T-LAK-cell-originated protein kinase (IC₅₀ = 2 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TOPK/PBK

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-29°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Kim, D., et al. 2012, Cancer Res.72, 3060.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service