Skip to Content
Merck
All Photos(1)

Key Documents

SML1654

Sigma-Aldrich

R-Ketorolac

≥95% (HPLC)

Synonym(s):

(+)-Ketorolac, (1R)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid;, (R)-(+)-ketorolac, (R)-Ketorolac

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$306.00
25 MG
$1,230.00

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$306.00
25 MG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C15H13NO3
CAS Number:
Molecular Weight:
255.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D +162 to +178°, c = 1 in methanol

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(O)[C@H]1C2=CC=C(C(C3=CC=CC=C3)=O)N2CC1

InChI

1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/t11-/m1/s1

InChI key

OZWKMVRBQXNZKK-LLVKDONJSA-N

Biochem/physiol Actions

Ketorolac ((rac)-5-benzoyl-1,2-3H-pyrrolo[1,2a] pyrrole1-carboxylic acid) is a non-steroidal anti-inflammatory drug (NSAID). It is used as a racemic mixture that contains a 1:1 ratio of the R(+) and S(−) stereoisomers. It is broadly used off-label as a parenteral analgesic in children.[1] Ketorolac serves as a non-narcotic analgesic. It prevents the synthesis of prostaglandins, peripheral to the central nervous system.[2]
R-Ketorolac is potent and selective Rho-family GTPases Cdc42 (cell division control protein 42) and Rac1 (Ras-related C3 botulinum toxin substrate 1) allosteric inhibitor that modulates downstream GTPase-dependent physiologic responses critical to tumor metastasis. R-ketorolac significantly inhibits ovarian cancer cell adhesion, migration, and invasion.
R-Ketorolac is potent and selective Rho-family GTPases Cdc42 and Rac1 allosteric inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuna Guo et al.
Molecular cancer therapeutics, 14(10), 2215-2227 (2015-07-25)
Cdc42 (cell division control protein 42) and Rac1 (Ras-related C3 botulinum toxin substrate 1) are attractive therapeutic targets in ovarian cancer based on established importance in tumor cell migration, adhesion, and invasion. Despite a predicted benefit, targeting GTPases has not
Tudor I Oprea et al.
Drug discovery today. Therapeutic strategies, 8(3-4), 61-69 (2012-03-01)
Academia and small business research units are poised to play an increasing role in drug discovery, with drug repurposing as one of the major areas of activity. Here we summarize project status for a number of drugs or classes of
The pharmacokinetics of ketorolac enantiomers following intramuscular administration of the racemate.
Hayball PJ, et al.
British Journal of Clinical Pharmacology, 37(1), 75-78 (1994)
Enantiomer?selective pharmacokinetics and metabolism of ketorolac in children.
Kauffman RE, et al.
Clinical Pharmacology and Therapeutics, 65(4), 382-388 (1999)
Tudor I Oprea et al.
PloS one, 10(11), e0142182-e0142182 (2015-11-13)
Rho family GTPases (including Rac, Rho and Cdc42) collectively control cell proliferation, adhesion and migration and are of interest as functional therapeutic targets in numerous epithelial cancers. Based on high throughput screening of the Prestwick Chemical Library® and cheminformatics we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service