Skip to Content
Merck
All Photos(1)

Documents

613551

Sigma-Aldrich

TMEM16A Inhibitor, T16Ainh-A01

The TMEM16A Inhibitor, T16Ainh-A01 controls the biological activity of TMEM16A.

Synonym(s):

TMEM16A Inhibitor, T16Ainh-A01, ANO1 Inhibitor, T16Ainh-A01, 2-(5-ethyl-4-hydroxy-6-methylpyrimidin-2-ylthio)-N-(4-(4-methoxyphenyl)thiazol-2-yl)acetamide, TMEM16A antagonist, T16Ainh-A01

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N4O3S2
CAS Number:
Molecular Weight:
416.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C19H20N4O3S2/c1-4-14-11(2)20-18(23-17(14)25)28-10-16(24)22-19-21-15(9-27-19)12-5-7-13(26-3)8-6-12/h5-9H,4,10H2,1-3H3,(H,20,23,25)(H,21,22,24)

InChI key

QSIYTNYMBWYHAA-UHFFFAOYSA-N

General description

An aminophenylthiazole that acts as an inhibitor of TMEM16A channel, a calcium-activated chloride channel (CaCC) (IC50 = 1.1 µM). Fully blocks CaCC current in salivary gland cells at 10 µM, but has very little effect in airway and intestinal cells, which is consistent with the observation of RNAi knock-down. Completely blocks chloride currents produced by TMEM16A Activator, Eact (Cat. No. 631547) at 10 µM. In addition, shown to block calcium-activated chloride channels in vascular smooth muscle cells, and relaxes murine and human blood vessels.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Davis, A.J., et al. 2012 Br. J. Pharmacol.,in press
Namkung, W., et al. 2011., J Biol Chem. in press.
Namkung, W., et al. 2011., FASEB J. in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylserine orchestrates Myomerger membrane insertions to drive myoblast fusion.
Gamage, et al.
Proceedings of the National Academy of Sciences of the USA, 119, e2202490119-e2202490119 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service