Skip to Content
Merck
All Photos(1)

Documents

5.89574

Sigma-Aldrich

1,2-Dimethoxyethane

99.5%, anhydrous

Synonym(s):

mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.22

grade

anhydrous

Quality Level

vapor density

3.1 (20 °C, vs air)

vapor pressure

48 mmHg ( 20 °C)

Assay

99.5%

autoignition temp.

396 °F

expl. lim.

10.4 %

refractive index

n20/D 1.379 (lit.)

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service