Skip to Content
Merck
All Photos(1)

Key Documents

8.08154

Sigma-Aldrich

Thionyl chloride

for synthesis

Synonym(s):

Thionyl chloride, Sulfurous acid dichloride, Sulfurous oxychloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2OS
CAS Number:
Molecular Weight:
118.97
MDL number:
UNSPSC Code:
12352307
EC Index Number:
231-748-8
NACRES:
NA.22
form:
liquid
Pricing and availability is not currently available.

vapor pressure

121 hPa ( 20 °C)

Quality Level

form

liquid

potency

324 mg/kg LD50, oral (Rat)

bp

76 °C/1013 hPa

mp

-104.5 °C

density

1.64 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(Cl)Cl

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

General description

Thionyl chloride is an inorganic acid chloride. It is mainly used as a chlorinating agent in organic synthesis for the production of organochlorine compounds.[1][2]

Application

Thionyl chloride can be used as a catalyst to synthesize:
  • 4-(sulfinylamino)-1,2,4-triazole by reacting with 4-amino-1,2,4-triazole.[1]
  • Isothiazolopyrimidines from aminouracils via cyclization reaction.[1]
  • 1,2,6-triazine S-oxides by reacting with diimines via condensation reactions in the presence of pyridine.[1]
  • 2-oxo-1,3,2-dioxathianes by condensation reaction with alkane-1,3-diols.[1]
  • Cyclic sulfites by reacting with nitroglycols.[1]
  • Microporous organic polymers by aldol self-condensation reaction of di and multiacetyl building blocks.[2]

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thionyl chloride-catalyzed preparation of microporous organic polymers through aldol condensation
Zhao Yan-Chao, et al.
Macromolecules, 44(16), 6382-6388 (2011)
Synthetic uses of thionyl chloride
El-Sakka IA and Hassan NA
Journal of Sulfur Chemistry, 26(1), 33-97 (2005)

Questions

  1. Do any of the Thionyl chlorides come under a SureSeal? If so, could I get the exact product #? I accidentally bought an ampule last time which was useless for my purposes.

    1 answer
    1. This product is packaged in glass bottles with a screw cap. See the links below to review products that are packaged with the Sure/Seal system:

      230464
      https://www.sigmaaldrich.com/product/sial/230464

      320536
      https://www.sigmaaldrich.com/product/sial/320536

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service