Skip to Content
Merck
All Photos(1)

Key Documents

5.42760

Millipore

DiluCult 2

Gravimetric Dilutor; Up to 5000g, suitable for sample preparation

Synonym(s):

Dilutor, Media dispenser

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121502
NACRES:
NA.84
Pricing and availability is not currently available.

manufacturer/tradename

DiluCult

application(s)

food and beverages
pathogen testing

sample preparation

shipped in

ambient

storage temp.

2-30°C

General description

Accurate microbiology results are highly reliant upon the quality of your sample preparation. Streamline your sample enrichment with our DILUCULT 2 gravimetric dilutors. These automated gravimetric dilutor instruments have two integrated pumps and are designed to facilitate work in a microbiological safety cabinet. They are low in height and have a removable drip tray. They operate with three modes of dispensing: a fast flow with lower accuracy, a standard flow with standard accuracy, and a slow flow for maximum accuracy. Choose your specific dilution factor on the user interface or switch to a dispensing mode for other applications. The DILUCULT 2 instrument has a robotic dispensing arm for higher safety and can be used for dilution of large pooling samples of up to 5000 g.

Application

DILUCULT 2 is a gravimetric dilutor instrument which is designed for automatic dilution of food testing samples with the necessary enrichment media to increase workflow efficiency. This instrument finds its application in pathogen and spoilage testing in food industry.

Features and Benefits

  • Automate repetitive workflow steps
  • Reduce hands-on time and error rates
  • DILUCULT 2 instrument is ideal for pooling samples
  • Add an additional pump for faster media dispensing

Components

Robotic arm, Bag holder, Protecting plate for nozzle, Removable drip tray, Nozzle holder and Tubing holder.

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DiluCult™ gravimetric dilutors automate food sample dilution, enhancing workflow efficiency in sample preparation.

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service