Skip to Content
Merck
All Photos(1)

Key Documents

528208

Sigma-Aldrich

Plasminogen Activator Inhibitor-1, Mutant

from Recombinant, ≥95% (SDS-PAGE), liquid, PAI-1 inhibitor, Calbiochem®

Synonym(s):

PAI-1, Mutant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Plasminogen Activator Inhibitor-1, Mutant, Human, Recombinant, The Plasminogen Activator Inhibitor-1, Mutant, Human, Recombinant controls the biological activity of Plasminogen Activator. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Highly purified preparation of an altered form of human PAI-1 containing four mutated amino acids. Mutant PAI-1 is virtually unable to go latent and is stable at elevated temperature and pH for extended periods of time (t½ = 145 hours at 37°C, pH 7.4). Inhibits uPA (Ki = 5.1 x 106 M-1 sec-1) and tPA (Ki = 7.9 x 105 M-1 sec-1).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Plasminogen Activator
Product does not compete with ATP.
Reversible: no
Target Ki: 5.1 x 106 M-1 sec-1, 7.9 x 105 M-1 sec-1 against uPA and tPA, respectively

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 50 mM sodium phosphate buffer, 1 mM EDTA, pH 6.6.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Berkenpas, M.B., et al. 1995. EMBO J. 14, 2969.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Bouarab et al.
Molecular psychiatry, 26(9), 4968-4981 (2021-01-30)
Moderate stress increases memory and facilitates adaptation. In contrast, intense stress can induce pathological memories as observed in post-traumatic stress disorders (PTSD). A shift in the balance between the expression of tPA and PAI-1 proteins is responsible for this transition.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service