Skip to Content
Merck
All Photos(1)

Documents

5.09704

Sigma-Aldrich

Cdc34 Inhibitor, CC0651

Synonym(s):

Cdc34 Inhibitor, CC0651, 4,5-Dideoxy-5-(3ʹ,5ʹ-dichloro-4-biphenylyl)-4-((methoxyacetyl)amino)-L-arabinonic acid, (2 R,3 S,4 S)-5-(3ʹ,5ʹ-Dichloro-(1,1ʹ-biphenyl)-4-yl)-2,3-dihydroxy-4-(2 methoxyacetamido)pentanoic acid, Cdc34A Inhibit

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21Cl2NO6
CAS Number:
Molecular Weight:
442.29
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

optical purity

ee: ≥98%

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

InChI

1S/C20H21Cl2NO6/c1-29-10-17(24)23-16(18(25)19(26)20(27)28)6-11-2-4-12(5-3-11)13-7-14(21)9-15(22)8-13/h2-5,7-9,16,18-19,25-26H,6,10H2,1H3,(H,23,24)(H,27,28)/t16-,18-,19+/m0/s1

InChI key

NTCBTNCWNRCBGX-YTQUADARSA-N

General description

A cell-permeable and aqueous soluble (~ 1 mg/mL or 2.26 mM at pH 7.4) biphenylylpentanoic acid that selectively inhibits human, but not yeast, E2 Ub-conjugating enzyme Cdc34- dependent substrates ubiquitinations in p27kip1-SCFSkp2, Sic1-SCFCdc4 (IC50 = 1.72 & 2.5 µM, respectively), Cyclin E-SCFFbw7, IκBα-SCFβTrCP ubiquitination assays and induces cellular p27kip1 and cyclin E accumulation (30 µM for 16 h in PC-3 cultures), displaying no affinity or inhibitory activity against other E1 (Uba1), E2 (Cdc34B/Ube2R2,Ube2G1, UbeU1), HECT E3 (SMURF 2, UbcH7), or RING E3 (Rnf168) tested. CC0651 is shown to target Ub-bound Cdc34 with a 14-fold higher affinity than the non-Ub-bound Cdc34 via an allosteric binding pocket at the periphery of the Cdc34-Ub interface. In addition to inhibit the reactivity of the Cdc34-Ub complex via direct binding, the CC0651-bound Cdc34-Ub complex itself is reported to exhibit higher affinity toward SCF E3 complex and thereby exhibit a dominant negative effect by effectively competing against functional Cdc34-Ub complex for SCF binding.
A cell-permeable biphenylylpentanoic acid compound that acts as a selective allosteric inhibitor of E2 ubiquitin-conjugating enzyme Cdc34 (IC50 = 2.5 µM in a Sic1-SCFCdc4 assay) without affecting the activities of E1 or E3 ligases. Reported to act by trapping a weak interaction between ubiquitin and the E2 donor ubiquitin-binding site and may serve as a molecular bridge between Cdc34A and ubiquitin. Binds to Cdc34ACAT alone with an EC50 value of 267 µM, but in the presence of ubiquitin, this affinity is significantly increased (EC50 = 19 µM). Does not disrupt the interaction of Cdc34A with the RING domain of Rbx1 and is suggested to even moderately stabilize this interaction. Shown to inhibit p27Kip1 ubiquitination (IC50 = 1.72 µM) and reduce PC-3 cancer cells proliferation (~30 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
E2 ubiquitin-conjugating enzyme Cdc34
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huang, H., et al. 2013. Nat. Chem. Biol.10, 156.

Ceccarelli, D.F., et al. 2011. Cell145, 1075.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service