Skip to Content
Merck
All Photos(1)

Key Documents

5.00519

Sigma-Aldrich

SKP2 E3 Ligase Inhibitor I, C1

Synonym(s):

SKP2 E3 Ligase Inhibitor I, C1, p21/Cip1/Waf1 Activator I; p27/Kip1 Activator I, S-phase Kinase-associated Protein 2 Inhibitor I; SCF SKP2 Inhibitor I, CRL1 SKP2 Inhibitor I, ( Z)-2-(4-Bromo-2-((4-oxo-3-(pyridin-3-ylmethyl)-2-thioxothiazolidin-5-ylidene)met

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13BrN2O4S2
CAS Number:
Molecular Weight:
465.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-brown

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

S1\C(=C/c3c(ccc(c3)Br)OCC(=O)O)\C(=O)N(C1=S)Cc2cnccc2

InChI

1S/C18H13BrN2O4S2/c19-13-3-4-14(25-10-16(22)23)12(6-13)7-15-17(24)21(18(26)27-15)9-11-2-1-5-20-8-11/h1-8H,9-10H2,(H,22,23)/b15-7-

InChI key

IYCJJVVXEHZJHE-CHHVJCJISA-N

General description

A cell-permeable rhodanine compound that upregulates cellular p21Cip1/Waf1 and p27Kip1 protein levels (10 µM for 16 h in 501 Mel cultures) by preventing Skp2-Cks1-mediated substrate recruitment without affecting the F-box E3 ubiquitin ligase Skp2 interaction with Cks1 or the SCF (Skp1-cullin1-F-box) complex. Shown to inhibit the proliferation of human breast cancer MCF-7 and melanoma 501 Mel cells (IC50 = 8 and 30 µM, respectively; 16 h incubation). Unlike SMIP004, C1 does not affect cellular Cyclin E/CDK2 kinase activity or Skp2 level.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
disrupts p27-Skp2 interaction
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Rico-Bautista, E., and Wolf, D.A. 2012. Chem. Biol.19, 1497.
Wu, L., et al. 2012. Chem. Biol.19, 1515.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service