Skip to Content
Merck
All Photos(2)

Key Documents

492040

Sigma-Aldrich

Nrf2 Activator

The Nrf2 Activator modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Nrf2 Activator, (E)-1-(2-Methoxyphenyl)-3-(2-(trifluoromethyl)phenyl)prop-2-en-1-one, Nuclear factor-erythroid 2 p45-related factor 2 Activator, 2-Trifluoromethyl-2ʹ-methoxychalone, Antioxidant Response Element Activator II, ARE Activator II, Antioxidant Response Element Activator II, ARE Activator II, (E)-1-(2-Methoxyphenyl)-3-(2-(trifluoromethyl)phenyl)prop-2-en-1-one, Nuclear factor-erythroid 2 p45-related factor 2 Activator, 2-Trifluoromethyl-2ʹ-methoxychalone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13F3O2
CAS Number:
Molecular Weight:
306.28
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

syrup

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL
ethanol: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(/C=C/C1=C(C=CC=C1)C(F)(F)F)C2=C(OC)C=CC=C2

General description

A cell-permeable chalcone that effectively activates NQO1 (NADPH-/quinone oxidoreductase 1) ARE- (antioxidant response element) mediated reporter transcription in human bronchial epithelial Beas-2B cells (7.0-fold of control; 20 µM compound) and induces Nrf2- (Nuclear factor erythroid 2 p45-related factor 2) regulated antioxidants and cytoprotective genes transcription, including GCLM (glutamate-cysteine ligase modifier subunit), NQO1, and HO1 (heme oxygenase-1), both in cultures ([compound] = 10 to 20 µM) in vitro and in mice (50 mg/kg p.o.) in vivo.
A cell-permeable chalcone that is shown to effectively activate NQO1 (NADPH/quinine oxidoreductase 1) ARE- (antioxidant response element) mediated reporter transcription in human bronchial epithelial Beas-2B cells (6.3- and 7.0-fold of control, respectively, with 10 and 20 µM compound) and induce Nrf2- (Nuclear factor erythroid 2 p45-related factor 2) regulated antioxidants and cytoprotective genes transcription, including GCLM (glutamate-cysteine ligase modifier subunit), NQO1, and HO1 (heme oxygenase-1), both in Beas-2B cultures (4-, 6.7-, and 178-fold, respectively, of basal NQO1/24 h, GCLM/6 h, and HO1/6 h mRNA level; [compound ] = 10 µM) in vitro and in mice (6- and 10-fold, respectively, of control GCLM and NQO1 mRNA level in small intestine 24 h after single p.o. dosage of 50 mg/kg) in vivo. The drug′s cellular target(s) and mechanism of action is unknown, but likely involves covalent thio modification of the target(s) by the compound′s electrophilic enone moiety.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Kumar, V., et al. 2011. J. Med. Chem.54, 4147.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dominga Manfredelli et al.
International journal of molecular sciences, 24(19) (2023-10-14)
The pathogenesis of coronavirus disease 2019 (COVID-19) is associated with a hyperinflammatory response. The mechanisms of SARS-CoV-2-induced inflammation are scantly known. Methylglyoxal (MG) is a glycolysis-derived byproduct endowed with a potent glycating action, leading to the formation of advanced glycation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service