Skip to Content
Merck
All Photos(4)

Key Documents

112941

Sigma-Aldrich

tert-Butylhydroquinone

97%

Synonym(s):

2-(1,1-Dimethylethyl)-1,4-benzenediol, NSC 4972

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$48.30
100 G
$55.00
500 G
$166.00

$48.30


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$48.30
100 G
$55.00
500 G
$166.00

About This Item

Linear Formula:
(CH3)3CC6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
166.22
Beilstein:
637923
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$48.30


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Quality Level

Assay

97%

autoignition temp.

855 °F

mp

127-129 °C (lit.)

SMILES string

CC(C)(C)c1cc(O)ccc1O

InChI

1S/C10H14O2/c1-10(2,3)8-6-7(11)4-5-9(8)12/h4-6,11-12H,1-3H3

InChI key

BGNXCDMCOKJUMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butylhydroquinone (TBHQ) is a phenolic antioxidant. TBHQ is frequently used as a food preservative. In low concentrations it shows cytoprotective qualities while at higher concentrations it exhibits cytotoxic behavior.[1]

Application

TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli.[2] Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ.[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon-Kyung Chung et al.
Journal of microbiology (Seoul, Korea), 46(3), 289-294 (2008-07-08)
Antimicrobial efficacy of ultra-high-pressure (UHP) can be enhanced by application of additional hurdles. The objective of this study was to systematically assess the enhancement in pressure lethality by TBHQ treatment, against barotolerant strains of Escherichia coli O157:H7 and Listeria monocytogenes.
Tert-butylhydroquinone-decorated graphene nanosheets and their enhanced capacitive behaviors
Wang HW, et al.
Chinese Science Bulletin, 56(20), 2092-9097 (2011)
Anna De Blasio et al.
Journal of cellular physiology, 235(2), 629-637 (2019-07-18)
The present study shows that nuclear factor erythroid 2-related factor 2 (NRF2) and miR-29b-1-5p are two opposite forces which could regulate the fate of MDA-MB-231 cells, the most studied triple-negative breast cancer (TNBC) cell line. We show that NRF2 activation
Saelooom Lee et al.
Free radical biology & medicine, 53(4), 807-816 (2012-06-12)
Radiotherapy and chemotherapeutic agents can effectively induce apoptosis through generation of reactive oxygen species (ROS). Cancer cells frequently express high levels of ROS-scavenging enzymes, which confer resistance to ROS-mediated cell death. Keap1 (Kelch-like ECH-associated protein 1) sequesters and promotes the
Tatsuya Ogawa et al.
Clinical, cosmetic and investigational dermatology, 13, 657-660 (2020-09-29)
The stratum corneum protects against the entry of pathogens, allergens, and irritants while preventing dehydration. The Kelch-like erythroid cell-derived protein with cap-n-collar homology-associated protein 1 (KEAP1)/NF-E2-related factor 2 (NRF2) system maintains skin barrier homeostasis. Aggregated evidence suggests that NRF2-mediated antioxidative

Questions

1–4 of 4 Questions  
  1. Do you have solution stability information on tert-Butylhydroquinone, Product 112941?

    1 answer
    1. Unfortunately, we do not have solution stability information on this product. We have not performed this type of testing in our labs, and we have no information from the chemical literature.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What type of applications can tert-Butylhydroquinone, Product 112941, be used for?

    1 answer
    1. While we do not perform specific use-testing for this product, literature reports that this product has been used as a stabilizer against autopolymerization of organic peroxide monomers.

      Helpful?

  4. What is tert-Butylhydroquinone, Product 112941, soluble in?

    1 answer
    1. The tert-Butylhydroquinone is soluble in ethanol (600 g/L at 25°C) and acetone (1120 g/L at 25°C). It is not expected to be soluble in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service