Skip to Content
Merck

Skip To

481981

Nifedipine

≥98% (UV), L-type Ca2+ channel bocker, solid

Synonym(s):

Nifedipine, L-Type Calcium Channel Blocker III

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H18N2O6
CAS Number:
Molecular Weight:
346.33
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (UV)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nifedipine, Relatively selective blocker of L-type Ca2+ channels.

description

Merck USA index - 14, 6528

Quality Segment

assay

≥98% (UV)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: soluble, ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

[N+](=O)([O-])c1c(cccc1)C2C(=C(NC(=C2C(=O)OC)C)C)C(=O)OC

InChI

1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3

InChI key

HYIMSNHJOBLJNT-UHFFFAOYSA-N

General description

Relatively selective blocker of L-type Ca2+ channels. One of the most extensively studied blockers of 1,4-dihydropyridine-type Ca2+ channels, and a pharmacologically-useful vasodilator. Induces apoptosis in human glioblastoma cells.

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Cell permeable: no
Primary Target
L-type Ca2+ channels

Preparation Note

Following reconstitution, refrigerate (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Reid, K., et al. 1997. J. Pharmacol. Exp. Ther. 283, 993.
Kondo, S., et al. 1995. J. Neurosurg.82, 469.
Antkiewicz-Michaluk, L., et al. 1994. Eur. J. Pharmacol. 254, 9.
Calabresi, P., et al. 1994. J. Neurosci. 14, 4871.
Akman, M.S., et al. 1993. Endocrinology132, 1286.
Cirillo, M., et al. 1993. Circ. Res.72, 847.
Young, R.J., et al. 1993. Biochim. Biophys. Acta1146, 81.
Janis, R., et al. 1987. Adv. Drug Res.16, 311.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N0750000PHR1290BP462
Nifedipine British Pharmacopoeia (BP) Reference Standard

BP462

Nifedipine

assay

≥98% (UV)

assay

-

assay

-

assay

-

form

solid

form

-

form

-

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

BP

Quality Level

100

Quality Level

-

Quality Level

300

Quality Level

-

storage temp.

10-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under Nifedipine


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
481981-250MG04055977183443

Questions

  1. Could you please provide the upper limit of saturation in DMSO for item 481981, Nifedipine?

    1 answer
    1. The highest test point found is that the substance is soluble in DMSO at a concentration of 50 mg/ml.

      Helpful?

Reviews

No rating value

Active Filters