Skip to Content
Merck
Nifedipine British Pharmacopoeia (BP) Reference Standard

Nifedipine

Nifedipine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester, Nifedipine
Empirical Formula (Hill Notation):
C17H18N2O6
CAS Number:
Molecular Weight:
346.33
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique