Skip to Content
Merck
All Photos(2)

Documents

443907

Millipore

D(-)Mannitol

Synonym(s):

D-Mannitol, Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

form

crystalline solid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0% sorbitol (HPLC)

color

white

mp

167-170 °C (lit.)

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effects of polyols at low concentration on the release of sweet aroma compounds in model soda beverages.: This study explores the influence of polyols, including D(-)Mannitol, on the release of sweet aroma compounds in soda beverages. The research demonstrates how low concentrations of D(-)Mannitol can enhance aroma profiles, providing valuable insights for the food and beverage industry. (Barba et al., 2024).

  • Blood-to-Testis Transport of Ribavirin Involves Carrier-Mediated Processes at the Blood-Testis Barrier.: This study investigates the transport mechanisms of Ribavirin across the blood-testis barrier, utilizing D(-)Mannitol as a key compound in the experimental setup. The findings contribute to the understanding of drug delivery systems targeting the blood-testis barrier. (Ito et al., 2024).

  • Staphylococcus hsinchuensis sp. nov., Isolated from Soymilk.: This study identifies a new species of Staphylococcus isolated from soymilk, with D(-)Mannitol playing a crucial role in the isolation and characterization process. The research enhances the understanding of microbial diversity in food products. (Wang et al., 2024).


Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service