Skip to Content
Merck
All Photos(2)

Documents

412510

Sigma-Aldrich

IRE1 Inhibitor I, STF-083010

The IRE1 Inhibitor I, STF-083010 controls the biological activity of IRE1. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

IRE1 Inhibitor I, STF-083010, ER-to-Nucleus Signaling 1 Inhibitor I, ERN1 Inhibitor I, Inositol-Reguiring Protein 1 Inhibitor I, STF083010, N-[(2-Hydroxynaphthalen-1-yl)methylidene]thiophene-2-sulfonamide, N-[(2-Hydroxy-1-naphthyl)methylene]-2-thiophenesulfonamide, N-[(2-Hydroxynaphthalen-1-yl)methylidene]thiophene-2-sulfonamide, N-[(2-Hydroxy-1-naphthyl)methylene]-2-thiophenesulfonamide, ER-to-Nucleus Signaling 1 Inhibitor I, ERN1 Inhibitor I, Inositol-Reguiring Protein 1 Inhibitor I, STF083010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO3S2
Molecular Weight:
317.38
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL, clear, yellow

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable salicylidene compound that is shown to directly target Ireα1 and disrupt Ire1-XBP1 (X-box binding protein) unfolded protein response (UPR) pathway in RPMI8226 multiple myeloma (MM) cells. Selectively inhibits ER stress-initiated endonuclease activity of Ire1 and blocks XPB1 mRNA splicing in MM cells at 60 µM and XBP1 activity in transgenic XBP1-luc mice (bortezomib 1mg/kg and 60 mg/kg of STF-083010, i.p.). Displays preferential cytotoxicity towards CD138+ cells over B, T and NK cells and antitumor properties.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Lerner, A.G., et al. 2012 Cell Metabolism16, 250.
Papandreou, I., et al. 2010. Blood117, )1311.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Demange et al.
eLife, 11 (2022-05-11)
Hundreds of cytotoxic natural or synthetic lipidic compounds contain chiral alkynylcarbinol motifs, but the mechanism of action of those potential therapeutic agents remains unknown. Using a genetic screen in haploid human cells, we discovered that the enantiospecific cytotoxicity of numerous
Jessica Proulx et al.
Frontiers in neuroscience, 16, 906651-906651 (2022-07-06)
Astrocytes are one of the most numerous glial cells in the central nervous system (CNS) and provide essential support to neurons to ensure CNS health and function. During a neuropathological challenge, such as during human immunodeficiency virus (HIV)-1 infection or
Jessica D Rosarda et al.
Nature communications, 14(1), 4119-4119 (2023-07-12)
The accumulation of atypical, cytotoxic 1-deoxysphingolipids (1-dSLs) has been linked to retinal diseases such as diabetic retinopathy and Macular Telangiectasia Type 2. However, the molecular mechanisms by which 1-dSLs induce toxicity in retinal cells remain poorly understood. Here, we integrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service